Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Brand: Thermo Scientific™ 75007621
Code : 51
Additional Details : Weight : 0.50000kg
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Conical | |
4-place Swing rotor with round bucket | |
15 mL |