missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorting Nexin 31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49084
This item is not returnable.
View return policy
Description
Sorting Nexin 31 Polyclonal antibody specifically detects Sorting Nexin 31 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Sorting Nexin 31 | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| MGC39715, sorting nexin 31, sorting nexin-31 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: WNEQLRRVFGNCLPPFPPKYYLAMTTAMADERRDQLEQYLQNVTMDPNVLRSDVFVEFLKLAQLNTFDIATKKAYLDIFLPNEQSIRIEIITSDTAERVLEVVSHKIGLCRELLGYFGLFLIRFGKEGKLSVVKKLADFELPYVSLGSSE | |
| 0.1 mL | |
| Signal Transduction | |
| 169166 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction