missing translation for 'onlineSavingsMsg'
Learn More

Sortin1, MedChemExpress

Product Code. 16481567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481567 50 mg 50mg
16445378 10 mg 10mg
16428097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481567 Supplier Medchem Express Supplier No. HY12827
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sortin1 is a chemical genetic-hit molecule that causes specific mislocalization of plant and yeast-soluble and membrane vacuolar markers.

Sortin1 is a chemical genetic-hit molecule that causes specific mislocalization of plant and yeast-soluble and membrane vacuolar markers.

Specifications

Chemical Name or Material Sortin1
CAS 503837-98-7
Color Red
Purity Grade Notes Research
Molecular Formula C26H19NO6
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (226.54 mM; Need ultrasonic)
SMILES O=C(C1=C(C)NC2=C(C(C3=C2C=CC=C3)=O)C1C4=CC=C(C5=CC=C(C(O)=O)C=C5)O4)OC
Molecular Weight (g/mol) 441.43
Formula Weight 441.43
Percent Purity 98.29%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.