missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sortilin Antibody (CL6528), Novus Biologicals™
Mouse Monoclonal Antibody
£292.00 - £433.00
Specifications
| Antigen | Sortilin |
|---|---|
| Clone | CL6528 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652560
|
Novus Biologicals
NBP2-76501-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203949
|
Novus Biologicals
NBP2-76501 |
100 μL |
£433.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sortilin Monoclonal specifically detects Sortilin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Sortilin | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Human, Mouse | |
| 100 kDa NT receptor, Glycoprotein 95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, sortilin, sortilin 1 | |
| SORT1 | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL6528 | |
| Monoclonal | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 6272 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LERNCEEKDYTIWLAHSTDPEDYEDGCILGYKEQFLRLRKSSVCQNGRDYVVTKQPSICLCSLEDFLCDFGYYRPENDSKCVEQPELKGHDLEFCLYGREEHLTTNGYRKIPGDKCQGGVNPVREVKD | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title