missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ SORT/TPL/G Calibration And Maintenance Buffers

Click to view available options
Quantity:
250 x 3 mL
Unit Size:
Each
Specifications
Specifications
| Chemical Name or Material | Sort/Tpl/Trace |
| Quantity | 250 x 3 mL |
| Includes | Buffer 4/7/10.01 (250mL each), 3 mol/L KCl (250mL), Pepsin cleaning solution (250mL) |
Safety and Handling
Product Identifier
- PEP/pH
- Warning
- H290-May be corrosive to metals.
- EUH208-Contains pepsin A. May produce an allergic reaction.
- P234-Keep only in original container.
- MIXTURE LIST-Contains : Pepsin A
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction