Learn More
SORT/TPL/G Calibration And Maintenance Buffers, Gel Electodes, WTW™
Product Code. 10599052
Click to view available options
Quantity:
250 x 2 mL
Unit Size:
Each
Product Code. 10599052
Brand: WTW 108823
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Calibration and maintenance are complete assortments for the calibration and maintenance of electrodes. The pepsin cleaning solution is used to remove protein containing precipitation from diaphragms, the 3 molar KCI as a storage or refill solution.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250 x 2 mL