missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SorLA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38177
This item is not returnable.
View return policy
Description
SorLA Polyclonal specifically detects SorLA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SorLA | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q92673 | |
| SORL1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GEKSTVFTIFGSNKENVHSWLILQVNATDALGVPCTENDYKLWSPSDERGNECLLGHKTVFKRRTPHATCFNGEDFDRPVVVSNCSCTREDYECDFGFKMS | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C11orf32, chromosome 11 open reading frame 32, FLJ21930, FLJ39258, gp250, Low-density lipoprotein receptor relative with 11 ligand-binding repeats, LR11LDLR relative with 11 ligand-binding repeats, LRP9, mosaic protein LR11, SorLA, SorLA-1SORLA, sortilin-related receptor, sortilin-related receptor, L(DLR class) A repeats containing, sortilin-related receptor, L(DLR class) A repeats-containing, Sorting protein-related receptor containing LDLR class A repeats | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6653 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction