missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORL1 Polyclonal Antibody

SORL1 Polyclonal Antibody
Brand: Invitrogen PA588941
This item is not returnable.
View return policy
Description
SORL1 Polyclonal Antibody for Western Blot
Specifications
| SORL1 | |
| Polyclonal | |
| Unconjugated | |
| SORL1 | |
| 2900010L19Rik; AI596264; AW261561; C11orf32; FLJ21930; FLJ39258; gp250; LDLR relative with 11 ligand-binding repeats; low-density lipoprotein receptor relative with 11 ligand-binding repeats; Lr11; LRP9; mosaic protein LR11; mSorLA; RGD619914; Sorl1; SorLA; SorLA-1; sortilin related receptor 1; sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; sortilin-related receptor, LDLR class A repeats-containing; sorting protein-related receptor containing LDLR class A repeats | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6653 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q92673 | |
| SORL1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 2065-2214 of human SORL1 (NP_0030961). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction