Learn More
Invitrogen™ SorCS3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA548023
Description
In direct ELISAs, approximately 40% cross-reactivity with recombinant human SorCS3 is observed, approximately 7% cross-reactivity with recombinant mouse (rm) SorCS1 is observed, and less than 2% cross-reactivity with rmSorCS2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
| SorCS3 | |
| Polyclonal | |
| Unconjugated | |
| SORCS3 | |
| 6330404A12Rik; AW045579; KIAA1059; SORCS; SORCS3; sortilin related VPS10 domain containing receptor 3; sortilin-related VPS10 domain containing receptor 3; VPS10 domain receptor protein SORCS 3 (SORCS3); VPS10 domain-containing receptor SorCS3 | |
| Goat | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66673 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8VI51 | |
| SORCS3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Ala131-Ser1122. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.