missing translation for 'onlineSavingsMsg'
Learn More

SorCS3 Antibody (339615) [DyLight 594], Novus Biologicals™

Product Code. 30732973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732973 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732973 Supplier Novus Biologicals Supplier No. FAB30671M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

SorCS3 Monoclonal antibody specifically detects SorCS3 in Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS3
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 339615
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias 6330404A12Rik, KIAA1059SORCS, sortilin-related VPS10 domain containing receptor 3, VPS10 domain receptor protein SORCS 3 (SORCS3), VPS10 domain-containing receptor SorCS3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SorCS3, Glu34-Ser1122, Accession # Q8VI51
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 22986
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.