missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SorCS2 Recombinant Mouse Monoclonal Antibody (K1E068_6E10)

Product Code. 30827783
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827783 100 μL
30825877 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827783 Supplier Invitrogen™ Supplier No. MA564498
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: EGGVDMQQSQ VQLQCPLTPP RGLQVSIQGE AVAVRPGEDV LFVVRQEQGD VLTTKYQVDL GDGFKAMYVN LTLTGEPIRH RYESPGIYRV SVRAENTAGH DEAVLFVQVN SPLQALYLEV VPVIGLNQEV NLTAVLLPLN PNLTVFYWWI GHSLQPLLSL DNSVTTRFSD TGDVRVTVQA ACGNSVLQDS RVLRVLDQFQ VMPLQFSKEL DAYNPNTPEW REDVGLVVTR LLSKETSVPQ ELLVTVVKPG LPTLADLYVL LPPPRPTRKR SLSSDKRLAA IQQVLNAQKI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Pig - 89%, Cynomolgus monkey - 97%.

This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SorCS2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K1E068_6E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene SORCS2
Gene Accession No. Q96PQ0
Gene Alias KIAA1329; mKIAA1329; N28137; SORCS2; SorCS2 104 kDa chain; SorCS2 122 kDa chain; SorCS2 18 kDa chain; sortilin related VPS10 domain containing receptor 2; sortilin-related VPS10 domain containing receptor 2; VPS10 domain receptor protein; VPS10 domain receptor protein SORCS 2; VPS10 domain-containing receptor SorCS2
Gene Symbols SORCS2
Host Species Mouse
Immunogen Recombinant human SORCS2 (aa 761-1050).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57537
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.