missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SorCS2 Monoclonal antibody specifically detects SorCS2 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
Specifications
| Antigen | SorCS2 |
| Applications | Western Blot, ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1D7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Sandwich ELISA |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Alias | KIAA1329VPS10 domain receptor protein, sortilin-related VPS10 domain containing receptor 2, VPS10 domain-containing receptor SorCS2 |
| Host Species | Mouse |
| Immunogen | SORCS2 (NP_065828.1, 802 a.a. ∽ 900 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VLRVLDQFQVMPLQFSKELDAYNPNTPEWREDVGLVVTRLLSKETSVPQELLVTVVKPGLPTLADLYVLLPPPRPTRKRSLSSDKRLAAIQQVLNAQKI |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?