missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SorCS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552883
This item is not returnable.
View return policy
Description
Immunogen sequence: EWRRDIGRVI KKSLVEATGV PGQHILVAVL PGLPTTAELF VLPYQDPAGE NKRSTDDLEQ ISELLIHTLN QNSVHFELKP GVRVLVHAAH LTAAPLVDLT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.
SORCS1 is one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein.
Specifications
| SorCS1 | |
| Polyclonal | |
| Unconjugated | |
| Sorcs1 | |
| FLJ41758; FLJ43475; FLJ44957; hSorCS; mSorCS; Sorcs; SORCS receptor 1; SORCS1; sortilin related VPS10 domain containing receptor 1; sortilin-related VPS10 domain containing receptor 1; VPS10 domain receptor protein SORCS 1; VPS10 domain-containing receptor SorCS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114815 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WY21 | |
| Sorcs1 | |
| Recombinant protein corresponding to Human SorCS1. Recombinant protein control fragment (Product #RP-90098). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction