missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORBS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24643-1-AP-150UL
This item is not returnable.
View return policy
Description
Arg and c-Abl represent the mammalian members of the Abelson family of non-receptor protein-tyrosine kinases. They interact with the Arg/Abl binding proteins via the SH3 domains present in the carboxy end of the latter group of proteins. ARGBP2 is the sorbin and SH3 domain containing 2 protein. It has three C-terminal SH3 domains and an N-terminal sorbin homology(SoHo) domain that interacts with lipid raft proteins. The subcellular localization of this protein in epithelial and cardiac muscle cells suggests that it functions as an adapter protein to assemble signaling complexes in stress fibers, and that it is a potential link between Abl family kinases and the actin cytoskeleton.Specifications
| SORBS2 | |
| Polyclonal | |
| Unconjugated | |
| SORBS2 | |
| Arg/Abl interacting protein 2, ARGBP2, KIAA0777, nArgBP2, PRO0618, Sorbin, SORBS2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 234214, 8470 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94875, Q3UTJ2 | |
| SORBS2 | |
| SORBS2 Fusion Protein Ag20041 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction