missing translation for 'onlineSavingsMsg'
Learn More

SORBS2 Antibody, Novus Biologicals™

Product Code. 18354269 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354269 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354269 Supplier Novus Biologicals Supplier No. H00008470B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

SORBS2 Polyclonal antibody specifically detects SORBS2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SORBS2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:2000
Formulation PBS (pH 7.4)
Gene Accession No. NP_066547.1
Gene Alias Arg/Abl interacting protein, Arg/Abl-interacting protein 2, ArgBP2, ARGBP2PRO0618, FLJ93447, KIAA0777Arg binding protein 2, Sorbin, sorbin and SH3 domain containing 2, sorbin and SH3 domain-containing protein 2
Host Species Mouse
Immunogen SORBS2 (NP_066547.1, 1 a.a. - 1100 a.a.) full-length human protein. MSYYQRPFSPSAYSLPASLNSSIVMQHGTSLDSTDTYPQHAQSLDGTTSSSIPLYRSSEEEKRVTVIKAPHYPGIGPVDESGIPTAIRTTVDRPKDWYKTMFKQIHMVHKPDDDTDMYNTPYTYNAGLYNPPYSAQSHPAAKTQTYRPLSKSHSDNSPNAFKDASSPVPPPHVPPPVPPLRPRDRSSTEKHDWDPPDRKVDTRKFRSEPRSIFEYEPGKSSILQHERPASLYQSSIDRSLERPMSSASMASDFRKRRKSEPAVGPPRGLGDQSASRTSPGRVDLPGSSTTLTKSFTSSSPSSPSRAKGGDDSKICPSLCSYSGLNGNPSSELDYCSTYRQHLDVPRDSPRAISFKNGWQMARQNAEIWSSTEETVSPKIKSRSCDDLLNDDCDSFPDPKVKSESMGSLLCEEDSKESCPMAWGSPYVPEVRSNGRSRIRHRSARNAPGFLKMYKKMHRINRKDLMNSEVICSVKSRILQYESEQQHKDLLRAWSQCSTEEVPRDMVPTRISEFEKLIQKSKSMPNLGDDMLSPVTLEPPQNGLCPKRRFSIEYLLEEENQSGPPARGRRGCQSNALVPIHIEVTSDEQPRAHVEFSDSDQDGVVSDHSDYIHLEGSSFCSESDFDHFSFTSSESFYGSSHHHHHHHHHHHRHLISSCKGRCPASYTRFTTMLKHERARHENTEEPRRQEMDPGLSKLAFLVSPVPFRRKKNSAPKKQTEKAKCKASVFEALDSALKDICDQIKAEKKRGSLPDNSILHRLISELLPDVPERNSSLRALRRSPLHQPLHPLPPDGAIHCPPYQNDCGRMPRSASFQDVDTANSSCHHQDRGGALQDRESPRSYSSTLTDMGRSAPRERRGTPEKEKLPAKAVYDFKAQTSKELSFKKGDTVYILRKIDQNWYEGEHHGRVGIFPISYVEKLTPPEKAQPARPPPPAQPGEIGEAIAKYNFNADTNVELSLRKGDRVILLKRVDQNWYEGKIPGTNRQGIFPVSYVEVVKKNTKGAEDYPDPPIPHSYSSDRIHSLSSNKPQRPVFTHENIQGGGEPFQALYNYTPRNEDELELRESDVIDVMEKCDDGWFVGTSRRTKFFGTFPGNYVKRL
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8470
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.