missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SORBS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13854-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in tyrosine phosphorylation of CBL by linking CBL to the insulin receptor. Required for insulin-stimulated glucose transport. Involved in formation of actin stress fibers and focal adhesions.Specifications
| SORBS1 | |
| Polyclonal | |
| Unconjugated | |
| SORBS1 | |
| c Cbl associated protein, CAP, Ponsin, R85FL, SH3 domain protein 5, SH3D5, SH3P12, SORB1, SORBS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10580, 20411 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q62417, Q9BX66 | |
| SORBS1 | |
| SORBS1 Fusion Protein Ag4828 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction