missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Mikromol™

Product Code. 17957234
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957234 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17957234 Supplier Mikromol Supplier No. MM1492.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Sorbitol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Sorbitol
CAS 50-70-4
Analyte or Component Names D-Sorbitol
Shipping Condition Room Temperature
Molecular Formula C6 H14 O6
Certifications/Compliance ISO 17034
Synonym D-Glucitol, Sorbitol, (-)-Sorbitol, 7B5697N, C*Sorbidex, C*Sorbidex 16100, C*Sorbidex P 16616, CSorbidex 16100, Cerestar P 16616, Cholaxine, Cystosol, D-(-)-Sorbitol, D-Sorbit 50M, D-Sorbol, Diakarmon, E 420, Esasorb, Foodol D 70, Glucarine, Glucarine (Sorbitol Syrup), Glucitol, Karion, Karion (Carbohydrate), Karion Instant, Kyowa Powder 50M, L-Gulitol, LTS P 50M, LTS-P 20M, Multitol, NSC 25944, Neosorb, Neosorb 20/60DC, Neosorb 70/02, Neosorb 70/02SB, Neosorb 70/20, Neosorb 70/70, Neosorb P 20/60, Neosorb P 60, Neosorb P 60W, Nivitin, P 60W, Parteck SI 150, Resulax, Sionit, Sionit K, Sionite, Sionon, Siosan, Sorbex M, Sorbex R, Sorbex Rp, Sorbex S, Sorbex X, Sorbidex 200, Sorbidex NC 16205, Sorbidex P 16616, Sorbidex S 16601, Sorbidex S 16603, Sorbierite, Sorbilande, Sorbilax, Sorbit, Erythritol Impurity B, Lactitol Monohydrate Impurity E, Maltitol Impurity A, Mannitol Impurity A, Lactitol Impurity E
InChI Formula InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
Quantity 250 mg
Formula Weight 182.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.