missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, LoGiCal

Product Code. 30459272
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30459272 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30459272

Brand: LoGiCal LGCFOR1492.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Sorbitol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Sorbitol
CAS 50-70-4
Analyte or Component Names D-Sorbitol
Shipping Condition Room Temperature
Molecular Formula C6 H14 O6
Certifications/Compliance ISO 9001
Synonym D-Glucitol, Sorbitol, (-)-Sorbitol, 7B5697N, C*Sorbidex, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W, P 60 W
InChI Formula InChI=1 S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12 H,1-2H2/t3-,4+,5-,6-/m1/s1
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO
IUPAC Name (2 R,3 R,4 R,5 S)-hexane-1,2,3,4,5,6-hexol
Industry Type Forensics
Molecular Weight (g/mol) 182.17
Quantity 10 mg
Formula Weight 182.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.