missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sorbitol Dehydrogenase Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518333
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.
Specifications
| Sorbitol Dehydrogenase | |
| Polyclonal | |
| Unconjugated | |
| SORD | |
| (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; HEL-S-95n; L-iditol 2-dehydrogenase; LOW QUALITY PROTEIN: sorbitol dehydrogenase; polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sdh1; Sdh-1; Sodh-1; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; SORD; SORD 1; SORD1; XDH; Xylitol dehydrogenase | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 20322 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q64442 | |
| SORD | |
| Synthetic peptide sequence (KIMLKCDPSDQNP) corresponding to the C-terminus amino acids of SORD (aa 345-357). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction