missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sorbitol Dehydrogenase Monoclonal Antibody for Western Blot, IP
Specifications
Specifications
| Antigen | Sorbitol Dehydrogenase |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | OTI2A10 |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SORD |
| Gene Accession No. | Q00796 |
| Gene Alias | (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; HEL-S-95n; L-iditol 2-dehydrogenase; LOW QUALITY PROTEIN: sorbitol dehydrogenase; polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sdh1; Sdh-1; Sodh-1; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; SORD; SORD 1; SORD1; XDH; Xylitol dehydrogenase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
