missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sorbitol Dehydrogenase Monoclonal Antibody (OTI1D5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524996
This item is not returnable.
View return policy
Description
Sorbitol Dehydrogenase Monoclonal Antibody for Western Blot, ICC/IF, Flow
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.
Specifications
| Sorbitol Dehydrogenase | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q00796 | |
| SORD | |
| Full length human recombinant protein of SORD produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| OTI1D5 | |
| Unconjugated | |
| SORD | |
| (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; HEL-S-95n; L-iditol 2-dehydrogenase; LOW QUALITY PROTEIN: sorbitol dehydrogenase; polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sdh1; Sdh-1; Sodh-1; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; SORD; SORD 1; SORD1; XDH; Xylitol dehydrogenase | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6652 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction