missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan sesquioleate

Product Code. 15454979
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
This item is not returnable. View return policy

Product Code. 15454979

Brand: Thermo Scientific Alfa Aesar J62648.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An emulsifier | CAS: 8007-43-0 | C42H76O7

Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel 'coated' after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel ‘coated‘ after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

Solubility
Insoluble in cold water, hot water. soluble in mineral oil and ethyl acetate.

Notes
Keep container tightly closed. Keep container in a cool, well-ventilated area and protect from moisture. 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8007-43-0
Molecular Formula C42H76O7
MDL Number MFCD00151568
Synonym Arlacel™ 83; Span™ 83

Specifications

Color Yellow
Physical Form Viscous Liquid
Density 0.989
Quantity 1 L
Flash Point 113°C (235°F)
Refractive Index 1.478
Solubility Information Insoluble in cold water,hot water. soluble in mineral oil and ethyl acetate.
Formula Weight 693.06
Chemical Name or Material Sorbitan sesquioleate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.