missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan monostearate

Product Code. 11367766
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367766 250 g 250g
11377766 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367766 Supplier Thermo Scientific Alfa Aesar Supplier No. L11435.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1338-41-6 | C24H46O6 | 430.63 g/mol

Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.

Solubility
Soluble in ethanol (50 mg/ml), isopropanol, mineral oil and vegetable oil. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
Synonym Sorbitan monooctadecanoate; Span™ 60
PubChem CID 23725022
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Melting Point 53°C to 57°C
Density 1
Quantity 250 g
Flash Point >149°C (300°F)
Merck Index 14,8724
Solubility Information Soluble in ethanol (50mg/ml),isopropanol,mineral oil and vegetable oil. Insoluble in water.
IUPAC Name [(2R)-2-[(3R,4S)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl] octadecanoate
Formula Weight 430.63
Chemical Name or Material Sorbitan monostearate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.