missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan monostearate

Product Code. p-7058372
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11367766

Brand: Thermo Scientific Alfa Aesar L11435.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1338-41-6 | C24H46O6 | 430.63 g/mol

Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan monostearate is used as an emulsifier and as a non-ionic surfactant. It is used in cosmetics, pharmaceuticals and food preparation.

Solubility
Soluble in ethanol (50 mg/ml), isopropanol, mineral oil and vegetable oil. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
Synonym Sorbitan monooctadecanoate; Span™ 60
PubChem CID 23725022
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

Melting Point 53°C to 57°C
Density 1
Quantity 250 g
Flash Point >149°C (300°F)
Merck Index 14,8724
Solubility Information Soluble in ethanol (50mg/ml),isopropanol,mineral oil and vegetable oil. Insoluble in water.
IUPAC Name [(2R)-2-[(3R,4S)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl] octadecanoate
Formula Weight 430.63
Chemical Name or Material Sorbitan monostearate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.