missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan Monostearate (Technical Grade), TRC

Product Code. 30003880
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003880 100 g 100g
30003631 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003880 Supplier Toronto Research Chemicals Supplier No. TRCS677005100G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.62
Synonym Ahco 909, Almax 3600, Anhydrosorbitol stearate, Arlacel 60, Armotan MS, Atmer 103, Crill 3, Crill K 3, Dehymul SMS, Disponil SMS, Drewsorb 60, Durtan 60, E 491, Emalex SPE 100, Emalex SPE 100S, Emasol 310, Emasol S 10, Emasol S 10V, Emasol S 10VHL, Emasol Super S 10F, Emsorb 2505, Estol 3715, Famodan MS, Grindsted SMS, Hodag SMS, Ionet S 60, Ionet S 60C, Ionet S 60V, Ionet S-60V, Liposorb S, Liposorb S 20, Lonzest SMS, MS 33, MS 33F, Montane 60, Montane 60PHA, Newcol 60, Nikkol SS 10, Nikkol SS 10M, Nikkol SS 10V, Nissan Nonion MP 30R, Nissan Nonion SP 60, Nissan Nonion SP 60R, Nissan Nonion SP 60RP, Nonion MP 30R, Nonion SP 60, Nonion SP 60R, Nonion SP 60RP, Poem 60V, Poem S 250, Poem S 60, Poem S 60V, Polycon 60, Polycon S 60K, Polycon S 80, Rheodol AS 10, Rheodol AS 10V, Rheodol SP-S 10, Rheodol SP-S 10V, Rheodol Super SP-S 10, Rikemal S 250, Rikemal S 300, S 300, S 60, S 7010, S-Maz 60k, SA 6638, SP 60R, SP 60RP, SP-S 10, SP-S 10V, SS 10, SS 10M, SS 10MV, SS 10V, Solman S 300, Solman S 301V, Sorbac 60, Sorbitan C, Sorbitan S, Sorbon S 60, Sorgen 50, Sorgen 50V, Span 55, Span 60, Span S 60, Span TS 1105, Sunsoft 61NN, Sunsoft 61S, Tego SMS, Sorbitan monooctadecanoate, Sorbitan Monostearate, Sorbitan Stearic Acid Monoester;
IUPAC Name [(2R)-2-[(2R,3R,4S)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl] octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC[C@@H](O)[C@H]1OC[C@H](O)[C@H]1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C24H46O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(27)29-19-21(26)24-23(28)20(25)18-30-24/h20-21,23-26,28H,2-19H2,1H3/t20-,21+,23+,24+/m0/s1
Formula Weight 430.3294
Chemical Name or Material Sorbitan Monostearate (Technical Grade)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.