missing translation for 'onlineSavingsMsg'
Learn More

Sorbic Acid, TRC

Product Code. 30133061
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133061 50 g 50g
17914085 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133061 Supplier Toronto Research Chemicals Supplier No. TRCS67689050G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6 H8 O2
Molecular Weight (g/mol) 112.13
Synonym Sorbic Acid, (2E,4E)-2,4-Hexadienoic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, (E,E)-2,4-Hexadienoic acid, α-trans-γ-trans-Sorbic acid, 2E,4E-Hexadienoic acid, E 200, NSC 35405, NSC 49103, NSC 50268, Panosorb, SO 215, Sorbistat, trans,trans-2,4-Hexadienoic acid, trans,trans-Sorbic acid
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+
Formula Weight 112.0524
Percent Purity >95%
Chemical Name or Material Sorbic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.