missing translation for 'onlineSavingsMsg'
Learn More

Soraprazan, MedChemExpress

Product Code. 16492576
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492576 25 mg 25mg
16415747 5 mg 5mg
16414316 100 mg 100mg
16413008 10 mg 10mg
16410649 1 mg 1mg
16409576 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16492576 Supplier Medchem Express Supplier No. HY100414
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Soraprazan (BYK61359) is a selective, reversible K-competitive inhibitor of the H,K-ATPase (Ki=6.4 nM), with an IC50 of 0.19 μM in gastric glands. Soraprazan binds to the H,K-ATPase with a Kd of 28.27 nM. Soraprazan shows immediate inhibition of acid secretion and is more than 2000-fold selective for H,K-ATPase over Na,K- and Ca-ATPases.

Soraprazan (BYK61359) is a selective, reversible K-competitive inhibitor of the H,K-ATPase (Ki=6.4 nM), with an IC50 of 0.19 μM in gastric glands. Soraprazan binds to the H,K-ATPase with a Kd of 28.27 nM. Soraprazan shows immediate inhibition of acid secretion and is more than 2000-fold selective for H,K-ATPase over Na,K- and Ca-ATPases.

Specifications

Chemical Name or Material Soraprazan
CAS 261944-46-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H25N3O3
Quantity 25 mg
Synonym BYK61359
Solubility Information DMSO : ≥ 150 mg/mL (408.23 mM)
SMILES O[C@@H]1[C@@H](C2=CC=CC=C2)NC3=C(C=CN4C3=NC(C)=C4C)[C@H]1OCCOC
Molecular Weight (g/mol) 367.44
Formula Weight 367.44
Percent Purity 99.69%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.