missing translation for 'onlineSavingsMsg'
Learn More

Sorafenib tosylate

Product Code. 16544504 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16544504

Brand: Thermo Scientific Acros 465710010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 475207-59-1 | C28H24ClF3N4O6S | 637.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 475207-59-1
Molecular Formula C28H24ClF3N4O6S
Molecular Weight (g/mol) 637.03
InChI Key IVDHYUQIDRJSTI-UHFFFAOYSA-N
IUPAC Name 4-[4-({[4-chloro-3-(trifluoromethyl)phenyl]carbamoyl}amino)phenoxy]-N-methylpyridine-2-carboxamide; 4-methylbenzene-1-sulfonic acid
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.CNC(=O)C1=CC(OC2=CC=C(NC(=O)NC3=CC=C(Cl)C(=C3)C(F)(F)F)C=C2)=CC=N1

Specifications

Quantity 1 g
Solubility Information Solubility aq. soln.: < 1 g/L (25°C). Other solubilities: < 1 g/L ethanol (25°C), 127 g/L dmso
Formula Weight 637.03
Chemical Name or Material Sorafenib tosylate
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H362-May cause harm to breast-fed children.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.