Learn More
Syngene™ Sony™ A6 Matte Paper
Product Code. 17776869 Shop All Syngene Products
Change view
Click to view available options
For Use With (Equipment):
A6 Digital Black and White Thermal Printer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17776869 A6 Digital Black and White Thermal Printer Each
1 options
Product Code. 17776869 Supplier Syngene™ Supplier No. UPP100HD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High density matte print media with proper width for use in the Sony™ A6 Digital Black and White Thermal Printer.
The UP-D898MD is a successor to the popular Sony™ UP-897 series of medical printers. Redesigned and upgraded, it offers several operational and ergonomic enhancements while retaining the same space-saving footprint and easy integration into medical carts.
  • Enhanced usability with enlarged LCD display panel and joystick for intuitive menu navigation
  • Delivers high quality black and white prints in under two seconds
  • Advanced thermal printing technology delivers near-photo quality results using widely available print media
  • Unique Sony™ technology allows paper to be cut in two directions – both up and down – as a lower cutter is newly included, along with the conventional upper cutter
  • Flexible printer installation in almost any location, as well as smooth printing operations
  • High quality thermal print engine can produce hard copy prints of still images captured for use in patient records and referrals
  • Type II matte paper
Specifications
Type Matte Paper
For Use With (Equipment) A6 Digital Black and White Thermal Printer
Dimensions (L x W) 20 m x 110 mm