missing translation for 'onlineSavingsMsg'
Learn More

Sonidegib, MedChemExpress

Product Code. 16483127
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483127

Brand: Medchem Express HY16582A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.

Sonidegib (Erismodegib) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.

Specifications

Chemical Name or Material Sonidegib
CAS 956697-53-3
Color Purple
Purity Grade Notes Research
Molecular Formula C26H26F3N3O3
Quantity 50 mg
Synonym Erismodegib LDE225 NVP-LDE225
Solubility Information DMSO : 50 mg/mL (102.99 mM; Need ultrasonic)
SMILES O=C(C1=C(C)C(C(C=C2)=CC=C2OC(F)(F)F)=CC=C1)NC3=CC=C(N=C3)N4C[C@@H](C)O[C@@H](C)C4
Molecular Weight (g/mol) 485.5
Formula Weight 485.5
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.