missing translation for 'onlineSavingsMsg'
Learn More

Sonidegib diphosphate, MedChemExpress

Product Code. p-200099299
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499627 5 mg 5mg
16473127 50 mg 50mg
16447088 10 mg 10mg
16477126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499627 Supplier Medchem Express Supplier No. HY16582

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sonidegib diphosphate (Erismodegib diphosphate) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.

Sonidegib diphosphate (Erismodegib diphosphate) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.

Specifications

Chemical Name or Material Sonidegib diphosphate
CAS 1218778-77-8
Color White
Purity Grade Notes Research
Molecular Formula C26H32F3N3O11P2
Quantity 5 mg
Synonym Erismodegib diphosphateLDE225 diphosphateNVP-LDE225 diphosphate
Solubility Information DMSO : 100 mg/mL (146.74 mM; Need ultrasonic) ∣H2O : 0.25 mg/mL (0.37 mM; Need ultrasonic)
SMILES O=C(C1=C(C)C(C(C=C2)=CC=C2OC(F)(F)F)=CC=C1)NC3=CC=C(N=C3)N4C[C@@H](C)O[C@@H](C)C4.O=P(O)(O)O.O=P(O)(O)O
Molecular Weight (g/mol) 681.49
Formula Weight 681.49
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.