missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sonidegib diphosphate, MedChemExpress
Description
Sonidegib diphosphate (Erismodegib diphosphate) is a potent and selective Smo antagonist with IC50 of 1.3 nM and 2.5 nM for mouse and human Smo in binding assay, respectively.
Specifications
Specifications
| Chemical Name or Material | Sonidegib diphosphate |
| CAS | 1218778-77-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H32F3N3O11P2 |
| Quantity | 5 mg |
| Synonym | Erismodegib diphosphateLDE225 diphosphateNVP-LDE225 diphosphate |
| Solubility Information | DMSO : 100 mg/mL (146.74 mM; Need ultrasonic) ∣H2O : 0.25 mg/mL (0.37 mM; Need ultrasonic) |
| SMILES | O=C(C1=C(C)C(C(C=C2)=CC=C2OC(F)(F)F)=CC=C1)NC3=CC=C(N=C3)N4C[C@@H](C)O[C@@H](C)C4.O=P(O)(O)O.O=P(O)(O)O |
| Molecular Weight (g/mol) | 681.49 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?