missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sonidegib
GREENER_CHOICE

Product Code. 18746926
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18746926

Brand: Tocris Bioscience™ 7826/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent Smoothened receptor antagonist

Sonidegib is a potent Smoothened (Smo) receptor antagonist (IC50 values are 1.3 nM and 2.5 nM at mouse and human receptors, respectively). Inhibits growth of a variety of tumor types including basal cell carcinoma, small cell lung cancer and medulloblastoma. Sonidegib inhibits Hh pathway in CD34+ chronic myeloid leukemia cells; it reduces both the number of cells and self-renewal ability. Brain penetrant and orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-N-[6-[(2R,6S)-2,6-Dimethyl-4-morpholinyl]-3-pyridinyl]-2-methyl-4'-(trifluoromethoxy)[1,1'-biphenyl]-3-carboxamide
CAS 956697-53-3
Quantity 50 mg
Target Smoothened
Molecular Formula C26H26F3N3O3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.