missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sonic Hedgehog/Shh Antibody (05), Alexa Fluor™ 488 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06169AF488
This item is not returnable.
View return policy
Description
Sonic Hedgehog/Shh Monoclonal antibody specifically detects Sonic Hedgehog/Shh in Human samples. It is validated for ELISA
Specifications
| Sonic Hedgehog/Shh | |
| Monoclonal | |
| Alexa Fluor 488 | |
| HHG1, HHG-1, HLP3, HPE3, MCOPCB5sonic hedgehog (Drosophila) homolog, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Sonic Hedgehog/Shh (Q15465; Met1-Gly197). | |
| 0.1 mL | |
| Biologically Active Proteins, Neuroscience, Sensory Systems, Stem Cell Signaling Pathway, Stem Cells, Vision | |
| 6469 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 05 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction