Learn More
SON Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28046-1-AP-20UL
This item is not returnable.
View return policy
Description
The SON antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SON. This antibody recognizes human, mouse antigen. The SON antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| SON | |
| 850 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| BASS1, C21orf50, DBP 5, DBP5, FLJ21099, FLJ33914, HSPC312, KIAA1019, NRE binding protein, NREBP, Protein DBP 5, Protein SON, SON, SON DNA binding protein, SON3 | |
| 20 μL | |
| 6651 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |