missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SON Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95174-0.02ml
This item is not returnable.
View return policy
Description
SON Polyclonal antibody specifically detects SON in Human samples. It is validated for Western Blot
Specifications
| SON | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BASS1C21orf50, Bax antagonist selected in saccharomyces 1, DBP5, DBP-5, FLJ33914, KIAA1019SON3, Negative regulatory element-binding protein, NREBPNRE-binding protein, Protein DBP-5, protein SON, SON DNA binding protein, SON DNA-binding protein, KIAA101910FLJ21099 | |
| A synthetic peptide corresponding to a sequence within amino acids 850-950 of human SON (NP_001278340.1). TMDSQMLATSSMDSQMLASGTMDSQMLASGTMDAQMLASGTMDAQMLASSTQDSAMLGSKSPDPYRLAQDPYRLAQDPYRLGHDPYRLGHDAYRLGQDPYR | |
| 0.02 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology | |
| 6651 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction