missing translation for 'onlineSavingsMsg'
Learn More

Somatostatin R4/SSTR4 Antibody (OTI2F11), DyLight 594, Novus Biologicals™

Product Code. 18427085 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427085 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427085 Supplier Novus Biologicals Supplier No. NBP274283DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Somatostatin R4/SSTR4 Monoclonal antibody specifically detects Somatostatin R4/SSTR4 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Somatostatin R4/SSTR4
Applications Western Blot
Classification Monoclonal
Clone OTI2F11
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias G-protein coupled receptor, somatostatin receptor 4, somatostatin receptor type 4, SS4R, SS-4-R, SS4-R
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues near C-terminus of human Somatostatin R4/SSTR4 (NP_001043).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 6754
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.