missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R4/SSTR4 Antibody (OTI2F11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74283CL1
This item is not returnable.
View return policy
Description
Somatostatin R4/SSTR4 Monoclonal antibody specifically detects Somatostatin R4/SSTR4 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Somatostatin R4/SSTR4 | |
| Monoclonal | |
| CoraFluor 1 | |
| G-protein coupled receptor, somatostatin receptor 4, somatostatin receptor type 4, SS4R, SS-4-R, SS4-R | |
| Synthetic peptide corresponding to residues near C-terminus of human SSTR4 (NP_001043). | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication, GPCR | |
| 6754 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| OTI2F11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction