missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R2/SSTR2 Antibody (7A9), Novus Biologicals™
Human Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Somatostatin R2/SSTR2 |
|---|---|
| Clone | 7A9 |
| Dilution | Flow Cytometry, ELISA |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30025636
|
Novus Biologicals
NBP3-26301-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30037486
|
Novus Biologicals
NBP3-26301-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Somatostatin R2/SSTR2 Monoclonal antibody specifically detects Somatostatin R2/SSTR2 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Somatostatin R2/SSTR2 | |
| Flow Cytometry, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| somatostatin receptor 2, somatostatin receptor type 2, SRIF-1, SS2R, SS-2-R, SS2-R | |
| Recombinant Human Somatostatin R2/SSTR2 protein [UniProt P30874] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 7A9 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Human | |
| Alzheimers Research, Cancer, Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 50% Glycerol, 0.01M PBS, PH 7.4 | |
| 6752 | |
| IgG1 | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title