missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Somatostatin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597200
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, SW620, MCF7, Mouse intestine, Rat stomach.
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.
Specifications
| Somatostatin | |
| Polyclonal | |
| Unconjugated | |
| SST | |
| Antrin; Growth hormone release-inhibiting factor; Neuronostatin; NST; preprosomatostatin; prepro-somatostatin; SMST; SOM; Somatostatin; somatostatin 28; somatostatin-14; Somatostatin-28; somatotropin release-inhibiting factor; SRIF; SS; SS-14; SS-28; Sst; SST-14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20604, 24797, 6750 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.62 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P60041, P60042, P61278 | |
| SST | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction