missing translation for 'onlineSavingsMsg'
Learn More

Somatostatin Antibody (CL15076) [Unconjugated], Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43653-25ul

 View more versions of this product

Product Code. 30279406

  • £285.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Somatostatin Monoclonal antibody specifically detects Somatostatin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Somatostatin
Monoclonal
1 mg/mL
Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500
Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28
This antibody was generated using a recombinant protein sequence of P61278, with the exact immunogen sequence remaining proprietary.
25 μL
Breast Cancer, Cancer, Stem Cell Markers
6750
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL15076
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.