missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin Antibody (CL15076) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44173
This item is not returnable.
View return policy
Description
Somatostatin Monoclonal antibody specifically detects Somatostatin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
| Somatostatin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry-Paraffin 1:200-1:500 | |
| Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28 | |
| This antibody was generated using a recombinant protein sequence of P61278, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 6750 | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry (Paraffin), Immunohistochemistry | |
| CL15076 | |
| Unconjugated | |
| Lyophilized in 5% Trehalose. | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
| Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction