missing translation for 'onlineSavingsMsg'
Learn More

Somatostatin Antibody (906552) [DyLight 488], Novus Biologicals™

Product Code. 30736322 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736322 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736322 Supplier Novus Biologicals Supplier No. FAB2358K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Somatostatin Monoclonal antibody specifically detects Somatostatin in Human,Mouse samples. It is validated for Immunohistochemistry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Somatostatin
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 906552
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Growth hormone release-inhibiting factor, SMST, somatostatin, somatostatin-14, somatostatin-28
Host Species Rat
Immunogen KLH-coupled human Somatostatin peptide, Accession # P61278
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6750
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.