Learn More
Ansell Edmont™ Solvex™ 37-675 Series Green Nitrile Immersion Gloves
Designed to deliver optimal results in wet or dry work environments where chemical resistance is crucial. Ansell Edmont™ Solvex™ 37-675 Series Green Nitrile Immersion Gloves are fully reusable with an unequaled abrasion resistance.
Brand:  Ansell Edmont™ 37-675_8
Product Code. 10060614
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Provides superb comfort
  • Sandpatch finish gives the glove a smooth surface that decreases costs thanks to fewer rejects of fragile parts
  • With cotton-flock liner and flexible nitrile film to enhance comfort
  • Ideal for chemical processing and preparation, oil and petrol refining, metal fabrication, agrochemicals and the printing industry