missing translation for 'onlineSavingsMsg'
Learn More

Solvent Violet 9 (Technical Grade), TRC

Product Code. 30424714
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424714 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424714 Supplier Toronto Research Chemicals Supplier No. TRCS67676310G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Solvent Violet 9 (Technical Grade)
CAS 467-63-0
Molecular Formula C25H31N3O
Quantity 10 g
Synonym Solvent Violet 9
InChI Formula InChI=1S/C25H31N3O/c1-26(2)22-13-7-19(8-14-22)25(29,20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6/h7-18,29H,1-6H3
SMILES CN(C)c1ccc(cc1)C(O)(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C
IUPAC Name tris[4-(dimethylamino)phenyl]methanol
Molecular Weight (g/mol) 389.54
Formula Weight 389.2467
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.