missing translation for 'onlineSavingsMsg'
Learn More

Solvent Red 52, TRC

Product Code. 30418058
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418058 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418058 Supplier Toronto Research Chemicals Supplier No. TRCS22439525MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Solvent Red 52
CAS 81-39-0
Molecular Formula C24 H18 N2 O2
Quantity 25 mg
Synonym 3-Methyl-6-[(4-methylphenyl)amino]-3H-naphtho[1,2,3-de]quinoline-2,7-dione, 2,3-Dihydro-3-methyl-2-oxo-6-p-toluidine-3-azabenzanthrone, 3-Methyl-6-(p-tolylamino)-2H-naphtho[1,2,3-de]quinoline-2,7(3H)-dione, 3-Methyl-6-p-toluidino-3H-dibenz[f,ij]isoquinoline-2,7-dione, Ahcoquinone Rubine Base, C.I. 68210, C.I. Solvent Red 52, Diaresin Red H 5B, Helio Oil Red R Pdr, KP Plast Red 5B, Kenawax Red Rubine 2RP, Macrolex Red 5B Macrolex Red 5B Gran, Oil Fast Red R Pdr, Optica Global PRT Red, Optica Global PRT Red Toner, Rubinol R Base, SR 52, Solvent Red 52, Sumiplast Red HL 5B, Transparent Red H 5B, Waxoline Rubine RA
InChI Formula InChI=1S/C24H18N2O2/c1-14-7-9-15(10-8-14)25-19-11-12-20-22-18(13-21(27)26(20)2)16-5-3-4-6-17(16)24(28)23(19)22/h3-13,25H,1-2H3
SMILES CN1C(=O)C=C2c3ccccc3C(=O)c4c(Nc5ccc(C)cc5)ccc1c24
IUPAC Name 14-methyl-10-(4-methylanilino)-14-azatetracyclo[7.7.1.02,7.013,17]heptadeca-1(16),2,4,6,9,11,13(17)-heptaene-8,15-dione
Molecular Weight (g/mol) 366.412
Formula Weight 366.137
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.