missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Solvent Blue 38

Product Code. 11428356
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428356 100 g 100g
11418356 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428356 Supplier Thermo Scientific Chemicals Supplier No. A15395.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol

Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.

Solubility
Soluble in water (partly), and methanol (1 mg/ml).

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1328-51-4
Molecular Formula C32H14CuN8Na2O6S2
Molecular Weight (g/mol) 780.16
MDL Number MFCD00071424
InChI Key DKBXPLYSDKSFEQ-UHFFFAOYSA-L
Synonym C.I. 74180; Direct Blue 86
PubChem CID 92030797
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=C(C=CC=C7S(=O)(=O)[O-])C(=N6)N=C8C9=C(C(=CC=C9)S(=O)(=O)[O-])C(=N8)N=C2[N-]3.[Na+].[Na+].[Cu+2]

Specifications

Melting Point ∼235°C (decomposition)
Solubility Information Soluble in water (partly),and methanol (1mg/ml).
Odor Odorless
Quantity 100 g
Chemical Name or Material Solvent Blue 38

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.