missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Solvent Blue 35

Product Code. 16471168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471168 5 g 5g
16408758 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471168 Supplier MedChemExpress Supplier No. HYD0516
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Solvent Blue 35 (Sudan Blue II; Oil Blue 35) is a dye used for colouring alcoholic and hydrocarbon based solvents. It is used for staining triglycerides in animal tissues.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 5 g
Shipping Condition Room temperature
Synonym Sudan Blue II ∣ Oil Blue 35
CAS 17354-14-2
Molecular Weight (g/mol) 350.45 g/mol
Solubility DMSO : 41.67 mg/mL (118.90 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.