missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Solvent Blue 35
Specifications
Specifications
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Product Type | Dye Reagents |
| Quantity | 10 g |
| Shipping Condition | Room temperature |
| Synonym | Sudan Blue II ∣ Oil Blue 35 |
| CAS | 17354-14-2 |
| Molecular Weight (g/mol) | 350.45 g/mol |
| Solubility | DMSO : 41.67 mg/mL (118.90 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble) |
| Target | HBV |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?