missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Solvent Blue 35

Product Code. p-200104504
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 16471168

Brand: MedChemExpress HYD0516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Solvent Blue 35 (Sudan Blue II; Oil Blue 35) is a dye used for colouring alcoholic and hydrocarbon based solvents. It is used for staining triglycerides in animal tissues.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 5 g
Shipping Condition Room temperature
Synonym Sudan Blue II ∣ Oil Blue 35
CAS 17354-14-2
Molecular Weight (g/mol) 350.45 g/mol
Solubility DMSO : 41.67 mg/mL (118.90 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Target HBV
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.